Methyl 2-bromo-3-chloro-6-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

32124-1G 32124-500MG 32124-5G
76940-080EA 706.56 USD
76940-080 76940-082 76940-084
Methyl 2-bromo-3-chloro-6-fluorobenzoate ≥95%
Methyl 2-bromo-3-chloro-6-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28141930
CAS Number: 1805210-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR