Methyl-6-chloro-5-methoxynicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-chloro-5-methoxynicotinate

32024-1G 32024-25G 32024-500MG 32024-5G
76940-066EA 129.43 USD
76940-066 76940-068 76940-070 76940-072
Methyl-6-chloro-5-methoxynicotinate ≥97% research grade
Methyl-6-chloro-5-methoxynicotinate
Formula: C₈H₈ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16659999
CAS Number: 915107-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR