tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate ≥95%

Supplier: AOBChem USA

Warning

31837-1G 31837-25G 31837-500MG 31837-5G
76940-028EA 126.25 USD
76940-028 76940-030 76940-032 76940-034
tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate ≥95%
tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate
Formula: C₁₆H₂₅BN₂O₄
MW: 320.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032217
CAS Number: 1095708-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR