Methyl 5-bromo-2-fluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

31753-1G 31753-500MG 31753-5G
76940-000EA 736.27 USD
76940-000 76940-002 76940-004
Methyl 5-bromo-2-fluoroisonicotinate ≥97%
Methyl 5-bromo-2-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698120
CAS Number: 1214375-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR