6-Bromo-2,3-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

31593-1G 31593-500MG 31593-5G
76939-980EA 468.92 USD
76939-980 76939-982 76939-984
6-Bromo-2,3-dichlorophenol ≥97%
6-Bromo-2,3-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MW: 241.897 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22055309
CAS Number: 1940-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR