6-Methoxy-2-naphthol ≥97%

Supplier: AOBChem USA

Warning

31519-25G 31519-5G
76939-968EA 410.57 USD
76939-968 76939-970
6-Methoxy-2-naphthol ≥97%
6-Methoxy-2-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00088677
CAS Number: 5111-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR