Ethyl 3-bromo-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

31437-1G 31437-25G 31437-500MG 31437-5G
76939-952EA 346.92 USD
76939-952 76939-954 76939-956 76939-958
Ethyl 3-bromo-5-methoxybenzoate ≥97%
Ethyl 3-bromo-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11042781
CAS Number: 1095274-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR