Ethyl 2,6-dimethoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

31326-1G 31326-25G 31326-500MG 31326-5G
76939-912EA 260.99 USD
76939-912 76939-914 76939-916 76939-918
Ethyl 2,6-dimethoxyisonicotinate ≥97%
Ethyl 2,6-dimethoxyisonicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22571563
CAS Number: 91013-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR