6-Bromo-8-fluoroquinoline ≥97%

Supplier: AOBChem USA

Warning

31323-25G 31323-5G
76939-900EA 474.23 USD
76939-900 76939-902
6-Bromo-8-fluoroquinoline ≥97%
6-Bromo-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053791
CAS Number: 220513-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR