Methyl 3-Chloro-6-fluoro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

31178-1G 31178-25G 31178-5G
76939-884EA 81.69 USD
76939-884 76939-886 76939-888
Methyl 3-Chloro-6-fluoro-2-methoxybenzoate ≥97%
Methyl 3-Chloro-6-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD29088271
CAS Number: 1879026-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR