Methyl 4-bromo-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

31162-1G 31162-25G 31162-500MG 31162-5G
76939-872EA 260.99 USD
76939-872 76939-874 76939-876 76939-878
Methyl 4-bromo-2,5-dimethylbenzoate ≥97%
Methyl 4-bromo-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18905462
CAS Number: 1195230-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR