5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

31103-1G 31103-25G 31103-5G
76939-840EA 96.55 USD
76939-840 76939-842 76939-844
5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester ≥97% research grade
5-Chloro-2-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)benzoic acid methyl ester
Formula: C₁₄H₁₈BClO₄
Storage Temperature: Refrigerator
MDL Number: MFCD16996244
CAS Number: 866625-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR