Methyl-4-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

30962-25G 30962-5G
76939-830EA 488.02 USD
76939-830 76939-832
Methyl-4-fluoro-3-methoxybenzoate ≥97%
Methyl-4-fluoro-3-methoxybenzoate
Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06203732
CAS Number: 74385-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR