Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

30907-1G 30907-25G 30907-500MG 30907-5G
76939-802EA 251.44 USD
76939-802 76939-804 76939-806 76939-808
Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Methyl 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula: C₁₅H₂₁BO₅
MW: 292.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494011
CAS Number: 478375-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR