Methyl 4-fluoro-2,6-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

30720-1G 30720-25G 30720-500MG 30720-5G
76939-740EA 118.83 USD
76939-740 76939-742 76939-744 76939-746
Methyl 4-fluoro-2,6-dimethylbenzoate ≥97%
Methyl 4-fluoro-2,6-dimethylbenzoate
Formula: C₁₀H₁₁FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06203803
CAS Number: 14659-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR