Methyl 2-chloro-5-fluoroisonicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-fluoro-4-(methoxycarbonyl)pyridine, Methyl 2-chloro-5-fluoropyridine-4-carboxylate

30544-1G 30544-25G 30544-500MG 30544-5G
76939-726EA 192.03 USD
76939-726 76939-728 76939-730 76939-732
Methyl 2-chloro-5-fluoroisonicotinate ≥97% research grade
Methyl 2-chloro-5-fluoroisonicotinate
Formula: C₇H₅ClFNO₂
MW: 189.572 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09909836
CAS Number: 876919-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR