Ethyl 2-ethoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

30530-1G 30530-25G 30530-5G
76939-712EA 88.06 USD
76939-712 76939-714 76939-716
Ethyl 2-ethoxynicotinate ≥97%
Ethyl 2-ethoxynicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18207652
CAS Number: 15441-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR