Methyl-5-hydroxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-hydroxy-2-methylbenzoate

30487-1G 30487-25G 30487-5G
76939-694EA 99.73 USD
76939-694 76939-696 76939-698
Methyl-5-hydroxy-2-methylbenzoate ≥97%
Methyl-5-hydroxy-2-methylbenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226199
CAS Number: 73505-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR