Methyl 2,6-difluoro-3-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

30426-1G 30426-25G 30426-500MG 30426-5G
76939-684EA 168.69 USD
76939-684 76939-686 76939-688 76939-690
Methyl 2,6-difluoro-3-iodobenzoate ≥97%
Methyl 2,6-difluoro-3-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₅F₂IO₂
MW: 298.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22573793
CAS Number: 501433-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR