N,N-di(propan-2-yl)-4-(trifluoromethyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

30373-25G 30373-5G
76939-672EA 204.76 USD
76939-672 76939-674
N,N-di(propan-2-yl)-4-(trifluoromethyl)benzamide ≥95%
N,N-di(propan-2-yl)-4-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD29088159
CAS Number: 79606-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR