Methyl-2,6-dimethoxypyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

30350-25G 30350-5G
76939-660EA 622.75 USD
76939-660 76939-662
Methyl-2,6-dimethoxypyridine-3-carboxylate ≥97%
Methyl-2,6-dimethoxypyridine-3-carboxylate
Formula: C₉H₁₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD15144586
CAS Number: 65515-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR