Methyl-2-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-fluorobenzoic acid methyl ester

30304-25G 30304-5G
76939-646EA 304.48 USD
76939-646 76939-648
Methyl-2-chloro-5-fluorobenzoate ≥97%
Methyl-2-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204207
CAS Number: 647020-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR