Methyl-2-bromo-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-bromo-3-(trifluoromethyl)benzoate

30140-1G 30140-25G 30140-500MG 30140-5G
76939-602EA 171.87 USD
76939-602 76939-604 76939-606 76939-608
Methyl-2-bromo-3-(trifluoromethyl)benzoate ≥97%
Methyl-2-bromo-3-(trifluoromethyl)benzoate
Formula: C₉H₆BrF₃O₂
MW: 283.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14697994
CAS Number: 1214362-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR