6-Bromo-3-isopropoxy-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

29985-1G 29985-25G 29985-500MG 29985-5G
76939-548EA 350.1 USD
76939-548 76939-550 76939-552 76939-554
6-Bromo-3-isopropoxy-2-methylpyridine ≥97%
6-Bromo-3-isopropoxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.102 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26393900
CAS Number: 1392466-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR