Methyl 4-bromo-3-chloro-2-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
29913-1G
29913-500MG
29913-5G
76939-526EA
891.16
USD
76939-526
76939-528
76939-530
Methyl 4-bromo-3-chloro-2-methylbenzoate ≥97%
Methyl 4-bromo-3-chloro-2-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrClO₂ MW: 263.516 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD23707507 CAS Number: 1427360-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...