Methyl 4-methylthiophene-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

29777-1G 29777-25G 29777-500MG 29777-5G
76939-510EA 213.25 USD
76939-510 76939-512 76939-514 76939-516
Methyl 4-methylthiophene-3-carboxylate ≥97%
Methyl 4-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD14698064
CAS Number: 61755-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR