Methyl 3-fluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

29694-25G 29694-5G
76939-502EA 537.88 USD
76939-502 76939-504
Methyl 3-fluoroisonicotinate ≥97%
Methyl 3-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045048
CAS Number: 876919-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR