Methyl 6-bromo-5-methoxynicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-bromo-5-methoxypyridine-3-carboxylate

29384-1G 29384-25G 29384-500MG 29384-5G
76939-440EA 213.25 USD
76939-440 76939-442 76939-444 76939-446
Methyl 6-bromo-5-methoxynicotinate ≥97% research grade
Methyl 6-bromo-5-methoxynicotinate
Formula: C₈H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD18257525
CAS Number: 1256810-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR