6-(Cyclopropylmethoxy)pyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

29119-1G 29119-25G 29119-500MG 29119-5G
76939-416EA 292.81 USD
76939-416 76939-418 76939-420 76939-422
6-(Cyclopropylmethoxy)pyridin-3-ol ≥97%
6-(Cyclopropylmethoxy)pyridin-3-ol

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28128936
CAS Number: 1224719-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR