N-(4-Bromophenyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

28985-1G 28985-25G 28985-500MG 28985-5G
76939-402EA 236.59 USD
76939-402 76939-404 76939-406 76939-408
N-(4-Bromophenyl)benzamide ≥97%
N-(4-Bromophenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD00017812
CAS Number: 7702-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR