Potassium 2-phenylethyltrifluoroborate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Potassium Trifluoro(phenethyl)borate, Potassium Trifluoro(2-phenylethyl)borate, Potassium Phenethyltrifluoroborate

28495-25G 28495-5G
76939-346EA 198.39 USD
76939-346 76939-348
Potassium 2-phenylethyltrifluoroborate ≥95%
Potassium 2-phenylethyltrifluoroborate

Precaución: for Research Use Only

Formula: C₈H₉BF₃K
Storage Temperature: Ambient
MDL Number: MFCD09039257
CAS Number: 329976-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR