Ethyl-6-bromo-1-benzothiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

28143-25G 28143-5G
76939-308EA 595.17 USD
76939-308 76939-310
Ethyl-6-bromo-1-benzothiophene-2-carboxylate ≥97%
Ethyl-6-bromo-1-benzothiophene-2-carboxylate
Formula: C₁₁H₉BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD11100112
CAS Number: 105191-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR