N-(3-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide ≥97%
Supplier: AOBChem USA
|
Warning
|
28104-1G
28104-25G
28104-500MG
28104-5G
76939-292EA
201.58
USD
76939-292
76939-294
76939-296
76939-298
N-(3-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide ≥97%
N-(3-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide
Precaución: for Research Use Only
Formula:
C₁₇H₂₅BClNO₃ MW: 337.649 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28142636 CAS Number: 2121512-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...