Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(Ethoxycarbonyl)pyridine-3-boronic acid pinacol ester

28085-1G 28085-25G 28085-500MG 28085-5G
76939-276EA 132.62 USD
76939-276 76939-278 76939-280 76939-282
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate ≥97%
Ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinate
Formula: C₁₄H₂₀BNO₄
MW: 277.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923436
CAS Number: 741709-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR