N-Cyano-N-phenyl-p-toluenesulfonamide ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
N-Cyano-4-methyl-N-phenylbenzenesulphonamide
28013-1G
28013-25G
28013-5G
76939-252EA
96.55
USD
76939-252
76939-254
76939-256
N-Cyano-N-phenyl-p-toluenesulfonamide ≥97%
N-Cyano-N-phenyl-p-toluenesulfonamide
Precaución: for Research Use Only
Formula:
C₁₄H₁₂N₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD00159358 CAS Number: 55305-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...