Methyl 4-chloro-5-fluoropyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

27896-1G 27896-500MG 27896-5G
76939-242EA 635.48 USD
76939-242 76939-244 76939-246
Methyl 4-chloro-5-fluoropyridine-3-carboxylate ≥97%
Methyl 4-chloro-5-fluoropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20925650
CAS Number: 1454913-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR