8-Bromo-6-methylimidazo[1,2-a]pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
27825-1G
27825-25G
27825-5G
76939-234EA
62.6
USD
76939-234
76939-236
76939-238
8-Bromo-6-methylimidazo[1,2-a]pyridine ≥97%
8-Bromo-6-methylimidazo[1,2-a]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂ MW: 211.0586 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09258770 CAS Number: 136117-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...