Methyl 6-bromo-2-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 6-bromo-2-methylnicotinate

27737-1G 27737-25G 27737-500MG 27737-5G
76939-204EA 343.74 USD
76939-204 76939-206 76939-208 76939-210
Methyl 6-bromo-2-methylpyridine-3-carboxylate ≥97%
Methyl 6-bromo-2-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611593
CAS Number: 1227562-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR