Methyl 2,6-dichloro-4-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

27661-1G 27661-500MG 27661-5G
76939-190EA 553.79 USD
76939-190 76939-192 76939-194
Methyl 2,6-dichloro-4-formylbenzoate ≥97%
Methyl 2,6-dichloro-4-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₃
Storage Temperature: Ambient
CAS Number: 1098620-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR