6-Bromo-2,4-dichlorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

27544-1G 27544-500MG 27544-5G
76939-172EA 561.22 USD
76939-172 76939-174 76939-176
6-Bromo-2,4-dichlorophenylboronic acid ≥95%
6-Bromo-2,4-dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrCl₂O₂
MW: 269.716 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088128
CAS Number: 2121514-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR