6-Bromo-2-chloronicotinic acid ≥95%

Supplier: AOBChem USA

Warning

27486-1G 27486-25G 27486-500MG 27486-5G
76939-134EA 199.45 USD
76939-134 76939-136 76939-138 76939-140
6-Bromo-2-chloronicotinic acid ≥95%
6-Bromo-2-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.451 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11857719
CAS Number: 1060815-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR