6-(1-Pyrrolidinyl)nicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

27358-1G 27358-25G 27358-500MG 27358-5G
76939-118EA 133.68 USD
76939-118 76939-120 76939-122 76939-124
6-(1-Pyrrolidinyl)nicotinic acid ≥97% research grade
6-(1-Pyrrolidinyl)nicotinic acid
Formula: C₁₀H₁₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD03791234
CAS Number: 210963-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR