Methyl 2-bromo-5-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 2-bromo-5-hydroxybenzenecarboxylate

27179-1G 27179-25G 27179-5G
76939-052EA 76.39 USD
76939-052 76939-054 76939-056
Methyl 2-bromo-5-hydroxybenzoate ≥97%
Methyl 2-bromo-5-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566797
CAS Number: 154607-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR