N-Benzyl-4-bromo-2-isopropylbenzamide ≥97%

Supplier: AOBChem USA

Warning

27092-1G 27092-25G 27092-500MG 27092-5G
76939-028EA 377.69 USD
76939-028 76939-030 76939-032 76939-034
N-Benzyl-4-bromo-2-isopropylbenzamide ≥97%
N-Benzyl-4-bromo-2-isopropylbenzamide

Precaución: for Research Use Only

Formula: C₁₇H₁₈BrNO
MW: 332.235 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32631798
CAS Number: 2364585-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR