6-Bromo-4-methylnicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-4-methylpyridine-3-carboxylic acid

27040-1G 27040-25G 27040-500MG 27040-5G
76939-012EA 350.1 USD
76939-012 76939-014 76939-016 76939-018
6-Bromo-4-methylnicotinic acid ≥97% research grade
6-Bromo-4-methylnicotinic acid
Formula: C₇H₆BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD13189138
CAS Number: 1060804-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR