6-Bromo-4-fluoro-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

26700-1G 26700-25G 26700-500MG 26700-5G
76938-958EA 290.69 USD
76938-958 76938-960 76938-962 76938-964
6-Bromo-4-fluoro-1-methyl-1H-indazole ≥97%
6-Bromo-4-fluoro-1-methyl-1H-indazole
Formula: C₈H₆BrFN₂
MW: 229.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756672
CAS Number: 1358574-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR