Methyl-3,5-dichloro-4-methoxybenzenecarboxylate ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3,5-dichloro-4-methoxybenzoate

26687-25G 26687-5G
76938-946EA 375.56 USD
76938-946 76938-948
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate ≥95% research grade
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Formula: C₉H₈Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00017189
CAS Number: 24295-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR