Methyl 6-fluoro-2-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

26502-1G 26502-25G 26502-500MG 26502-5G
76938-904EA 144.29 USD
76938-904 76938-906 76938-908 76938-910
Methyl 6-fluoro-2-methylnicotinate ≥97%
Methyl 6-fluoro-2-methylnicotinate
Formula: C₈H₈FNO₂
MW: 169.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611614
CAS Number: 1227595-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR