6-Quinoxaline​boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

26496-1G 26496-25G 26496-5G
76938-894EA 102.91 USD
76938-894 76938-896 76938-898
6-Quinoxaline​boronic acid pinacol ester ≥97%
6-Quinoxaline​boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BN₂O₂
MW: 256.1080 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11054040
CAS Number: 1167418-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR