7-Methyl-1H-indazole-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 7-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

26494-1G 26494-25G 26494-500MG 26494-5G
76938-884EA 292.81 USD
76938-884 76938-886 76938-888 76938-890
7-Methyl-1H-indazole-4-boronic acid pinacol ester ≥97%
7-Methyl-1H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27756661
CAS Number: 1186334-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR